Learn More
Cole-Parmer N015.5CM Grade No. 1 Qualitative Filter Paper; 5.5 cm dia.; 100/pk
Supplier:  Cole-Parmer 8104804
Encompass_Preferred
Ideal for clarifying and removing precipitates and preparation for qualitative analysis. These papers are made of 100% alpha cotton cellulose, with 0 to 12 pH tolerance. Maximum temperature is 248F (120C).
Catalog No. NC1529716
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.