Learn More
Essendant FILTTERFAUCSBRITA WATER
Supplier:  Essendant CLO42201CT
Encompass_Preferred
Brita Faucet Mount System filters out odors and impurities for healthier great tasting filtered water on tap From the world s 1 brand in water filtration this Elite Advanced faucet mount uses Advanced Brita Carbon-block technology is certified to reduce chlorine (taste and odor) and reduces 60plus contaminants including 99pct of lead This Brita filter system replaces 1 800 bottles a year This certified Brita water filter system has three spray options and includes a filter indicator to tell when it s time to change approximately three times a year Does not fit pull-out or spray style faucets White 4 8 wx2 26 dx5 28 h Water Filters Type- Faucet Filter Quantity- 4/Carton
Catalog No. 50-000-22160
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.