Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ fimC (Escherichia coli) Recombinant Protein

Catalog No. 89944589 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-589 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-944-589 Supplier Abnova™ Supplier No. P4835
Add to Cart
Add to Cart

Used for Func, SDS-PAGE

Sequence: MGSSHHHHHHSSGLVPRGSHMGVALGATRVIYPAGQKQEQLAVTNNDENSTYLIQSWVENADGVKDGRFIVTPPLFAMKGKKENTLRILDATNNQLPQDRESLFWMNVKAIPSMDKSKLTENTLQLAIISRIKLYYRPAKLALPPDQAAEKLRFRRSANSLTLINPTPYYLTVTELNAGTRVLENALVPPMGESTVKLPSDAGSNITYRTINDYGALTPKMTGVME

Specifications

Accession Number NP_418736
Concentration 1 mg/mL
For Use With (Application) Functional Study, SDS-PAGE
Formulation Liquid
Gene ID (Entrez) 948843
Molecular Weight (g/mol) 25kDa
Name fimC (Escherichia coli) Recombinant protein
Preparation Method Escherichia coli expression system
Purification Method Conventional Chromatography
Quality Control Testing 15% SDS-PAGE Stained with Coomassie Blue
Quantity 100 μg
Immunogen MGSSHHHHHHSSGLVPRGSHMGVALGATRVIYPAGQKQEQLAVTNNDENSTYLIQSWVENADGVKDGRFIVTPPLFAMKGKKENTLRILDATNNQLPQDRESLFWMNVKAIPSMDKSKLTENTLQLAIISRIKLYYRPAKLALPPDQAAEKLRFRRSANSLTLINPTPYYLTVTELNAGTRVLENALVPPMGESTVKLPSDAGSNITYRTINDYGALTPKMTGVME
Storage Requirements Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ECK4307/JW4279
Common Name fimC
Gene Symbol fimC
Species E. coli
Protein Tag His
Expression System Escherichia coli expression system
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.