Learn More
Biomatik Corporation Fimh Polyclonal Antibody
Supplier:  Biomatik Corporation CAC1171750UL
Encompass_Preferred
Fimh Polyclonal Antibody is a Escherichia coli (strain K12) Polyclonal Antibody to fimH. The Fimh Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Escherichia coli (strain K12). Immunogen: Recombinant Escherichia Coli Type 1 Fimbrin D-Mannose Specific Adhesin (Fimh) (22-300Aa). Applications: ELISA, WB. Target Synonyms: fimH. Accession Number: P08191. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. NC2112391
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.