Learn More
Chemglass Life Sciences Findenser Mini, 275mm, B24 Outer and Inner Joints
SDP
Supplier:  Chemglass Life Sciences CG1218F124
Encompass_Preferred
Findenser™ comprises of an internal glass condenser and an external, finned aluminum heat sink jacket. The area between the internal glass condenser and the finned aluminum jacket is filled with a small amount of water that is permanently sealed. The internal design of the glass condenser provides an increased internal surface area compared to traditional condensers. The larger surface area improves the heat transfer capacity of the condenser. The Findenser replaces the need for water cooled condensers in over 95% of all common chemistry applications
Catalog No. NC1629248
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.