Learn More
Fine Science Tools Adson Forceps, Stainless Steel, Straight, 12 cm
SDP
Supplier:  Fine Science Tools 1102712
Encompass
Tip Shape: Straight/ Tips: 1x2 Teeth/ Tip Dimensions: 1.5 x 0.8mm/ Alloy / Material: Stainless Steel/ Length: 12cm
Catalog No. NC9924848