Learn More
Epredia™ Fine Pore Cassettes
Catalog No. B851735OR Shop All Epredia Products
Change view
Click to view available options
Color:
Beige
Blue
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
B851735OR Orange
B851735BE Beige
B851735BL Blue
B851735GN Green
B851735LA Lavender
B851735LG Light Green
B851735LI Lilac
B851735PK Pink
B851735TN Tan
B851735WH White
B851735YW Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. B851735OR Supplier Epredia™ Supplier No. B851735OR
Add to Cart
Add to Cart
For very small biopsy tissue samples, cassettes have a front-edge lid hinge with attached lids that are available in 12 different color varieties
Printing
  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
  • Precision engineering for solid connection to microtomes
Archiving
  • Cassette surface texture optimizes scannability for archiving
Specifications
Pore Style Fine Pore
Packaging Type Loose Packed
Quantity 1000/Cs.
Closure Included Yes
Closure Type Attached Lid, Front Hinge
PrintMate Compatible No
Color Orange
Product Type Cassette