Learn More
Thermo Scientific™ Finger Tite Connectors
Catalog No. 290ST130 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Female Nut
Catalog No. Type
29-0ST-130 Female Nut
1 options
Catalog No. 29-0ST-130 Supplier Thermo Scientific™ Supplier No. 290ST130
Add to Cart
Add to Cart
Ensure reliable, leak-free connections for optimal GC performance using these Finger Tite Connectors.
Ensure reliable, leak-free connections for optimal GC performance using Thermo Scientific™ Finger Tite Connectors. The perfect metal-to-metal seal eliminates contamination and produces lower air/water background noise compared to conventional graphite/polymer ferrules. They are easy to handle even with a hot injector/detector and do not require the use of tools. Unlike other ferrules, they do not require retightening after installation.
Recommended Use
  • Thermo Scientific™ and Agilent™ GC instruments
  • Connectors for SSL and MS systems
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) TRACE GC Ultra and FOCUS GC Instruments
Description Female Nut
Type Female Nut
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.