Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Impact Gloves with Thumb Padding & Detachable Wrist Support

Catalog No. 19031880 Shop All Impacto Protective Products Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Large
Hand:
Specific
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Hand
19-031880 Small Specific
19-031873 Large Specific
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-031880 Supplier Impacto™ Supplier No. 70410120020
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Detachable suede leather wrist support helps to prevent over flexion and over extension. Additional padding in the thumb webbing prevents cumulative trauma that can occur from repetitive gripping.

  • Nylon fabric ensures breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm and thumb web area protects from impact
  • Fingerless style for tactile feedback and mobility
  • Suede leather cover offers abrasion protection
  • Detachable leather wrist support prevents over flexion and over extension to help prevent RSI
  • For protection from hand tools and power tools
  • Ideal for applications in construction, assembly, manufacturing, maintenance. and warehousing

Specifications

Back Material Nylon Lycra™
Cuff Material Suede Leather
Color Blue, Gray
Hand Specific
Material Suede
Product Type Anti-impact Gloves
Size Small
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Color Gray
Series 704-10
Length (English) 7 in.
Length (Metric) 18 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.