Learn More
MAC-MOD Analytical PEEK Fingertight Fitting (10 pack) – suitable for connection of all Partisil and Partisphere columns to all 1/16”o.d. tubing types – slip free to 6000psi
SDP
Supplier:  MAC-MOD Analytical HI050X
Encompass_Preferred
Suitable for connection of all Partisil and Partisphere columns to all 1/16¿o.d. tubing types ¿ slip free to 6000psi.
Catalog No. 50-304-805
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.