Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Fingertip Bandage BMD Unit

Catalog No. 19091116 Shop All Medique Products
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-091-116 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-091-116 Supplier Medique Supplier No. 6501BMD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medique™ Fingertip Bandage BMD Unit is a refill for ANSI-2015 Class A First Aid Kit.

  • Blue
  • Metal Detectable
  • Woven Fingertip (10 count)

Specifications

Product Type Bandage
For Use With (Application) ANSI-2015 Class A First Aid Kit Refill
Adhesive Yes
Color Blue
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.