Learn More
Medchemexpress LLC HY-11063 100mg Medchemexpress, Fingolimod CAS:162359-55-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11063100MG
Encompass_Preferred
Medchemexpress, HY-11063 100mg Fingolimod CAS:162359-55-9 Fingolimod (FTY720 free base) is a sphingosine 1-phosphate (S1P) antagonist with an IC50 of 0.033 nM in K562 and NK cells. Fingolimod also is a pak1 activator, a immunosuppressant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0784
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.