Description
Dose liquids from reagent bottles with Thermo Scientific™ Finnpipette™ Dispensers.
Features:
- Excellent chemical and thermal resistance
- Fully autoclavable†
- Exact self-locking system in the volume setting
- Dispensing head cap prevents dripping
- Ergonomic design prevents work fatigue
- Threaded suction hose for bubble-free suction
Note:
Limitations include HF, liquids that attack Halar, FEP or Hastelloy suspensions because solid particles might block the valves.
Specifications
Specifications
| Product Type | Analog Bottletop Dispenser |
| Certifications/Compliance | Individual calibration certificate with serial number |
| Description | Range: 5-30mL, ±180µL |
| Channels | 1 |
| Volume (Metric) | 5.0 to 30.0 mL |
| Precision | 60 μL |
Note: Not for use on hot plates.