Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Finnpipette™ Dispenser

Catalog No. 14487205 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
0.2 to 1.0 mL
0.4 to 2.0 mL
1.0 to 5.0 mL
10.0 to 60.0 mL
2.0 to 10.0 mL
5.0 to 30.0 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-487-205 10.0 to 60.0 mL
14-487-200 0.2 to 1.0 mL
14-487-201 0.4 to 2.0 mL
14-487-202 1.0 to 5.0 mL
14-487-203 2.0 to 10.0 mL
14-487-204 5.0 to 30.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-487-205 Supplier Thermo Scientific™ Supplier No. 4421170
  • $2,300.00
    $1,602.65 / Each of 1

    Save $697.35

    30% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dose liquids from reagent bottles with dispensers that offer superior chemical and thermal resistance, and an ergonomic design that prevents work fatigue.

Dose liquids from reagent bottles with Thermo Scientific™ Finnpipette™ Dispensers.

Features:

  • Excellent chemical and thermal resistance
  • Fully autoclavable†
  • Exact self-locking system in the volume setting
  • Dispensing head cap prevents dripping
  • Ergonomic design prevents work fatigue
  • Threaded suction hose for bubble-free suction

Note:

Limitations include HF, liquids that attack Halar, FEP or Hastelloy suspensions because solid particles might block the valves.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Analog Bottletop Dispenser
Certifications/Compliance Individual calibration certificate with serial number
Description Range: 10-60mL, ±360µL
Channels 1
Volume (Metric) 10.0 to 60.0 mL
Precision 120 μL

Note: Not for use on hot plates.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.