Learn More
Thermo Scientific™ Finnpipette™ F1 Fixed Volume Single Channel Pipettes
GREENER_CHOICE
Catalog No. 14386327N
Change view
Click to view available options
Volume (Metric):
10 μL
10,000 μL
100 μL
1000 μL
20 μL
200 μL
2000 μL
25 μL
250 μL
3000 μL
50 μL
500 μL
5000 μL
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-386-327N 50 μL
14-386-325N 10 μL
14-386-326N 25 μL
14-386-328N 100 μL
14-386-329N 250 μL
14-386-330N 500 μL
14-386-331N 1000 μL
14-386-332N 2000 μL
14-386-333N 3000 μL
14-386-334N 5000 μL
14-386-335N 10,000 μL
14-386-296N 20 μL
14-386-297N 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 14-386-327N Supplier Thermo Scientific™ Supplier No. 4651040N
Add to Cart
Add to Cart
Choose these pipettes for set protocols. They deliver a light, smooth pipetting action and an adjustable finger rest to maximize your comfort.
Great for set protocols and QA environments, Thermo Scientific™ Finnpipette™ F1 Fixed Volume Pipettes deliver a light, smooth pipetting action and an adjustable finger rest to maximize your comfort.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F1: Everything you want in your everyday, go-to pipette
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μl models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
CE marked
The Finnpipette F1 Pipette together with compatible Finntip tips is CE marked in accordance with IVD Directive in Europe.
5 Year Warranty
With convenient online registration, and may vary by country.
Learn more about the complete Finnpipette family >
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips Finntip 250, Filter 100, Flex 200, Flex Filter 100
Description Finnpipette F1 Fixed Volume
Number of Channels 1
Type Fixed Volume Pipette
Volume (Metric) 50 μL