Learn More
Thermo Scientific™ Finnpipette™ Multistepper Pipette
Catalog No. 21377371
Change view
Click to view available options
Volume (Metric):
1250 μL
Catalog No. Volume (Metric)
21-377-371 1250 μL
1 options
Catalog No. 21-377-371 Supplier Thermo Scientific™ Supplier No. 2206590
Reduce the risk of error and increase productivity in immunoassay applications with this multistepper pipette, designed especially for work with microplates and featuring simultaneous 8-channel dispensing.
Reduce the risk of error and increase productivity in immunoassay applications with the Thermo Scientific™ Finnpipette™ Multistepper Pipette. This lightweight, easy-to-use repeater pipette provides simultaneous dispensing in eight channels. Designed especially for work with microplates, the Finnpipette Multistepper helps speed up routine laboratory procedures and is easy to use — simply attach, fill, and prime the module, then dial the selected volume.
Design
  • Universal handle also fits the seven single-channel Thermo Scientific™ Finntip™ Steppers
  • Lightweight construction minimizes hand fatigue during repetitive dispensing
  • Separate tip ejector for safe and effortless tip disposal
  • Wide finger rest minimizes operator hand strain
  • Module rotates a full 360°, optimizing efficiency for both right or left-handed users
Compatible with:
Accepts Finntip Multistepper 1500μL tips. They are extra long, with more air space between the liquid and tip cone to prevent contamination.
TRUSTED_SUSTAINABILITY
Specifications
Description 8-Channel Module only
For Use With Microplate
Number of Channels 8
Type Multistepper Pipette
Volume (Metric) 1250 μL