Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Finnpipette™ Novus GLP Kits
GREENER_CHOICE

Catalog No. p-4530136
Change view
Click to view available options
Volume (Metric):
1 to 1000 μL
10 to 10,000 μL
10 to 1000 μL
100 to 5000 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-386-104 10 to 1000 μL
14-386-105 100 to 5000 μL
14-386-106 1 to 1000 μL
14-386-107 10 to 10,000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-386-104 Supplier Thermo Scientific™ Supplier No. 4700470
Add to Cart
Edge
Add to Cart

Access everything needed for pipetting in these all-inclusive GLP kits featuring ergonomic and easy-to-use pipetters.

Access everything needed for pipetting in these all-inclusive Thermo Scientific™ Finnpipette™ Novus GLP kits. The kits cover the volume range you need with four different options to choose from, and include Finnpipette™ Novus™ pipetters, appropriate stands and sample tips.

Novus GLP Kit 1 includes:

  • Finnpipette Novus 10-100μL
  • Finnpipette Novus 100-1000μL
  • Two Finnpipette Novus stands
  • One rack of Finntip Flex 200 and Flex 1000
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Pipetting Guide
  • Finnpipette Novus brochure

Novus GLP Kit 2 includes:

  • Finnpipette Novus 100-1000μL
  • Finnpipette Novus 0.5-5mL
  • Two Finnpipette Novus stands
  • One rack of Finntip 5ml and Flex 1000
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Finnpipette Novus brochure
  • Pipetting Guide

Novus GLP Kit 3 includes:

  • Novus pipette, 1-10μL, micro
  • Novus pipette, 10-100μL
  • Novus pipette, 100-1000μL
  • Three Finnpipette Novus pipette stands
  • One rack of Flex 10, Flex 200 and Flex 1000
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Pipetting Guide
  • Novus brochure

Novus GLP Kit 4 includes:

  • Novus pipette, 10-100μL
  • Novus pipette, 100-1000μL
  • Novus pipette, 1-10mL
  • Three Finnpipette Novus pipette stands
  • One rack of Flex 200, Flex 1000 and Finntip 10ml
  • Reagent Reservoirs demo pack
  • Finntip Sample Pack
  • Pipetting Guide
  • Novus brochure
TRUSTED_SUSTAINABILITY

Specifications

Color Assorted
Model Novus
Number of Channels 1
Product Line Novus
Type GLP Kit
Volume (Metric) 10 to 1000 μL
Includes Novus 100 and 1000 μL single channel pipettes, two Novus stands, two sample Finntip racks
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.