Learn More
Thermo Scientific™ Finnpipette™ Stands
GSA_VAGREENER_CHOICE
Catalog No. 1438721
Change view
Click to view available options
Product Type:
Carousel Stand with Cover
Pipette Stand
Holds:
Electronic Pipette
Manual Pipettes (3)
Manual Pipettes (6)
Manual Pipettes (6) or Electronic Pipettes (3)
Multi-channel Manual Pipette
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Holds
14-387-21 Pipette Stand Electronic Pipette
14-245-160 Pipette Stand Manual Pipettes (6)
14-387-14 Pipette Stand Manual Pipettes (6)
14-387-15 Pipette Stand Manual Pipettes (3)
21-377-002 Carousel Stand with Cover Manual Pipettes (6) or Electronic Pipettes (3)
14-245-161 Pipette Stand Multi-channel Manual Pipette
21-377-125 - -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 14-387-21 Supplier Thermo Scientific™ Supplier No. 9420360
Add to Cart
Add to Cart
Provide safe and convenient storage for manual and electronic pipettes using these stands, with options for single stand, F-stand, multi-stand, multichannel and carousel.
Provide safe and convenient storage for manual and electronic pipettes using Thermo Scientific™ Finnpipette™ Stands. Available options include options for single stand, F-stand, multi-stand, multichannel and carousel.
Finnpipette F-Stand
  • Accommodates six pipettes
  • Sturdy crossbar structure with slot for each pipette for secure storage
  • Suitable for both single channel and multichannel models
Finnpipette Multichannel Stand
  • Multi-position stand
  • Can hold any multichannel Finnpipette
  • Pipette can be stored in upright or horizontal position
  • Small footprint
Finnpipette Carousel Stand
  • Rotating, portable
  • Transparent lid protects pipettes from contamination
  • Shipped with three adapters for electronic Finnpipette Novus pipettes and five adapters for manual Finnpipette pipettes
Finnpipette Novus Stand
  • For convenient storage of electronic Finnpipette Novus pipette
  • Accommodates both single and multichannel Finnpipette Novus models with ease
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Color White
Holds Electronic Pipette
Product Type Pipette Stand