Learn More
Invitrogen™ FIP1L1 Polyclonal Antibody

Description
Immunogen sequence: KANSSVGKWQ DRYGRAESPD LRRLPGAIDV IGQTITISRV EGRRRANENS NIQVLSERSA TEVDNNFSKP PPFFPPGAPP T Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
Specifications
Specifications
| Antigen | FIP1L1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FIP1L1 |
| Gene Accession No. | Q6UN15 |
| Gene Alias | 1300019H17Rik; DKFZp586K0717; Factor interacting with PAP; factor interacting with PAPOLA and CPSF1; FIP1; FIP1 like 1; FIP1 like 1 (S. cerevisiae); FIP1 like 1b (S. cerevisiae); FIP1L1; FIP1L1 cleavage and polyadenylation specific factor subunit; fip1l1b; FIP1-like 1 protein; FLJ33619; hFip1; Pre-mRNA 3'-end-processing factor FIP1; pre-mRNA 3'-end-processing factor FIP1; LOW QUALITY PROTEIN: pre-mRNA 3'-end-processing factor FIP1; rearranged in hypereosinophilia; RHE; Rje; wu:fd36f11; zgc:103421 |
| Gene Symbols | FIP1L1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.