Learn More
TARGETMOL CHEMICALS INC FIPAMEZOLE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T24064
Encompass_Preferred
Also available in 50 mg 100 mg and bulk. Please contact Fisher for quotes. Fipamezole is a potent alpha2 adrenergic receptor antagonist that may be useful for studying autonomic dysfunction in Parkinson's disease. purity: 99%
Catalog No. 50-258-0833
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.