Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Firefly luciferase Monoclonal Antibody (Luci17)
GREENER_CHOICE

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA112556

Catalog No. 01-674-342


Only null left
Add to Cart

Description

Description

MA1-12556 detects Luciferase in C. elegans and Drosophilia melanogaster tissue, human fibroblasts, mouse macrophages, kidney, liver and cortex tissues, as well as NIH-3T3, Jurkat and BHR21 cell lines with little to no background signal.

Luciferase produces green light with the wavelength of 562 nm. Luciferase catalyzes the reaction: D-firefly luciferin + O2 + ATP = firefly oxyluciferin + CO2 + AMP + diphosphate + light.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Firefly luciferase
Monoclonal
1 mg/mL
PBS with 0.08% sodium azide
Mouse
Protein A/G
RUO
Luciferase
Antibody
IgG1
Immunohistochemistry (Frozen), Western Blot
Luci17
Unconjugated
Firefly Luciferase; Genji firefly; Green Luciferase; Japanese firefly; luciferin 4-monooxygenase; Photinus pyralis; photinus pyralis luciferase
Luciferase protein isolated from Photinus pyralis.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.