Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Firefly Luciferase Recombinant Protein
SDP

Catalog No. p-7112741 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB5964820UG 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB5964820UG Supplier Novus Biologicals™ Supplier No. NBP25964820UG
Only null left
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-311 of Renilla reniformis Luciferase The Recombinant Invertebrate Luciferase His (N-Term) Protein is derived from E. coli. The Recombinant Invertebrate Luciferase His (N-Term) Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.

Specifications

For Use With (Application) Bioactivity
Formulation 20mM Tris-HCl (pH8.0) containing 1mM DTT, 10% glycerol
Molecular Weight (g/mol) 38.5 kDa
Name Firefly Luciferase Recombinant Protein
Quantity 20 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias ec 1.13.12.7, Fluc
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Firefly
Recombinant Recombinant
Purity or Quality Grade >95% by PAGE
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.