Learn More
MSA™ FireHawk™ M7 Air Mask Accessory, Nitrile O-Ring
Nitrile O-ring prevents leakage and is a critical component of the SCBA.
Supplier:  MSA™ 60709
 View more versions of this product
Catalog No. MSA60709
Description
Description
  • Replacement part for SCBA air system
  • Prevents air leakage
  • The O-ring should be inspected regularly and thoroughly checked based on usage
  • Found on the high pressure hose that connects to the air cylinder
Specifications
Specifications
M7 Air Mask
0.07 in.
O-ring
60709
2.512 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.