Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

International Enviroguard™ FirmGrip Shoe Covers

Catalog No. 19472479 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Large
X-Large
Material:
CPE
Proprietary Laminated Non-woven Fabric
Color:
Blue
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Material Color
19-472-479 Large Proprietary Laminated Non-woven Fabric Blue
19-472-478 Large Proprietary Laminated Non-woven Fabric White
19-472-480 X-Large CPE White
19-472-481 X-Large CPE Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-472-479 Supplier International Enviroguard™ Supplier No. 3701B
Add to Cart
Add to Cart

Shoe covers provide an extra sticky firm and secure grip to reduce the risk for slips, trips, and falls.

FirmGrip non-skid shoe covers are designed for tough working environments. Not only do they protect from particulates, but they also safeguard against light liquid splashes. Using a multi-layer fabric and textured finish, these durable shoe covers offer the traction to get things done without slipping. A strong elastic closure keeps shoe covers tightly in place

  • Textured surface for a firm grip in slick or wet environments
  • Strong elastic closure provides a secure fit
  • Size Large fits men's sizes 6 to 11
  • Size Extra Large fits men's sizes 12 to 16
  • Common applications include: Healthcare, Construction, Realtors, Movers, Delivery Services, Food Processing, Cleanrooms, Installers, and more
  • Can be worn in indoor and outdoor environments

Specifications

Size Large
Material Proprietary Laminated Non-woven Fabric
Latex Free Yes
Color Blue
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Non-skid
For Use With (Application) Construction, Realtors, Movers, Delivery Services, Food Processing, Cleanrooms, Installers
Packaging Quantity 300/Cs.
Resistance Dry Particulates, Splash
Product Type Controlled Environments Shoe Covers
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.