Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique First-Aid Handbook

Catalog No. 19090674 Shop All Medique Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-090-674 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-090-674 Supplier Medique Supplier No. 71401

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

General guide to basic first aid measures.

  • Pocket-sized
  • For quick reference

Specifications

Product Type First Aid Handbook
For Use With (Application) General guide to basic first aid measures
Description For quick reference
Format Pocket-sized
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.