Learn More
Essendant FIRST AIDEYEWSH STATION
Supplier:  Essendant FND320020000000
Encompass_Preferred
Fifteen-minute 100pct sterile eye wash station Alarm sounds upon activation and completion of 15-minute flush Easy to replace cartridge which has a life of 24 months (sold separately) An integrated waste containment prevents spills and provides easy clean up Constant flow rate meets ANSI Z358 1-2014 standard for primary eyewash station Mounting Details- Wall Depth- 34 75 Height- 17 5 Width- 15 5
Catalog No. 50-000-31684
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.