Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva First-Strand cDNA Synthesis Kit

Catalog No. 45001414 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
55 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-414 55 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-414 Supplier Cytiva Supplier No. 27926101
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Bulk first-strand cDNA reaction mixes (Cloned murine reverse transcriptase, RNase inhibitor, RNase/DNase-free BSA, dATP, dCTP, dGTP, and dTTP in aqueous buffer), mRNA standard, DTT solution, RNase-free water, pd(N)6 primer, and Not I-(dT)19 bifunctional primer

For synthesis of single-stranded cDNA from mRNA templates prior to amplification by PCR

  • Preassembled bulk reaction mixes save time and minimize potential for pipetting errors
  • Primers are not included in the preassembled reaction mixes, allowing use of a primer of choice
  • Two primers are included with the kit
    • pd(N)6 primer (random hexamer) is used to prepare libraries for screening with antibodies, to increase the representation of 5'-ends of mRNAs having significant secondary structure, or to copy mRNAs lacking a poly(A) tail
    • Not I-(dT)18 bifunctional primer can be used to prime selectively on mRNA having a poly(A) tail and can also be used as a PCR primer following first-strand synthesis
    • The Not I restriction site, located at the 5'-end of the primer, is particularly useful for subsequent directional cloning of cDNA and PCR products

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -15°C to -30°C
Description Designed to generate full-length first-strand cDNA from mRNA templates. Following first-strand cDNA synthesis, the sample can be used directly for in vitro amplification using PCR or for second-strand synthesis using the Gubler-Hoffman method.
For Use With (Application) RT-PCR
Includes Bulk first strand cDNA reaction mixes, mRNA standard, DTT solution, RNase-free Water, pd(N)6 primer and NotI-(dT)18 bifunctional primer
No. of Reactions 55
Product Type First-Strand cDNA Synthesis Kit
Quantity 55 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.