Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences First Strand cDNA Synthesis Kit
SCI_ED

Catalog No. p-200080570
Change view
Click to view available options
No. of Reactions:
30
150
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
S03140 150
S03141 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S03140 Supplier G-Biosciences Supplier No. 7861977
Only null left
Add to Cart
Add to Cart

First strand cDNA synthesis kit contains Reverse Transcriptase (RT), recombinant RNase inhibitor, dNTPs and an optimized buffer with protein stabilizers. It is used for gene expression analysis.

  • First strand cDNA synthesis kit is in a 5x master mix form which contains Reverse Transcriptase (RT), recombinant RNase inhibitor, dNTPs and an optimized buffer with protein stabilizers.
  • G-biosciences RT is a recombinant Moloney Murine Leukemia Virus (MMLV) reverse transcriptase with reduced RNase H activity.
  • This RT is an RNA-directed DNA polymerase which lacks 3' - 5' exonuclease activity and can produce full-length cDNA from long RNA templates.

Specifications

Detection Method qPCR, RT-PCR
Form Liquid
GC-Rich PCR Performance High
PCR Method qPCR, 1-step RT-qPCR, 2-step RT-qPCR, Standard PCR, 1-step RT-PCR
Reaction Speed Fast or Standard
Label or Dye Unconjugated, Unlabeled, No ROX, Biotin and Other Hapten, Biotin
Product Type Synthesis Kit
For Use With (Application) Gene Expression, Genotyping, Microbial Detection, cDNA Synthesis, Genetic Disease Research
No. of Reactions 150
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.