Learn More
AdipoGen Fisetin
SDP
Supplier:  AdipoGen CDXF0336M500
Encompass_Preferred
Chemical. CAS 528-48-3. Formula C15H10O6. MW 286.24. Synthetic. One of the major polyphenolic flavonoids found in various fruits and vegetables such as apples, grapes, persimmons, strawberries, cucumbers and onions. Exerts numerous beneficial biological activities, including antibacterial, antifungal, antioxidant, anti-inflammatory, antiangiogenic, antiproliferative, proapoptotic, cell cycle arrest, autophagy inducing, hypolipidemic, neuroprotective and antitumor effects. Modulates multiple signaling pathways such as blocking of the phosphatidylinositol-3-kinase/protein kinase B/mammalian target of rapamycin PI3K/Akt/mTOR and p38, mitogen-activated protein kinases MAPK-dependent nuclear factor kappa-light-chain-enhancer of activated B cells NF-kappaB signaling, Wnt and adrogen receptor signaling, which playing a central role in various cellular processes contributing to the malignancy. Binds to beta-tubulin and disrupts microtubule dynamics.
Catalog No. 50-256-0856
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.