Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Fisetin
GREENER_CHOICE

Catalog No. 501650R
Change view
Click to view available options
:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
501650R 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 501650R Supplier Tocris Bioscience™ Supplier No. 5016/50
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Naturally occuring flavonoid and antioxidant; neuroprotective

Fisetin is a naturally occuring flavonoid and antioxidant. Inhibits PI 3-K, Akt, mTOR and Cdk6. Displays antiproliferative activity in prostate cancer cells. Shown to activate ERK; exhibits neuroprotective activity in Huntington's disease models. Also a DNMT1 inhibitor.

Specifications

Quantity 50 mg
Product Type Fisetin
Purity >98%
Molecular Weight (g/mol) 286.24
Inhibitors Fisetin

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.