Learn More
3M™ Fish Protein Rapid Kit L25FSH, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S26913 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Fish Protein Rapid Kit L25FSH, Sold by AquaPhoenix Scientific
Catalog No.
S26913 Fish Protein Rapid Kit L25FSH, Sold by AquaPhoenix Scientific
1 options
Catalog No. S26913 Supplier 3M™ Supplier No. L25FSH
Add to Cart
Add to Cart
For screening for the presence of fish proteins in clean-in-place (CIP) final rinse water, environmental swab samples, food ingredients and processed food products
  • Results visible at 11 ± 1 min. after application of sample
  • Rapid, qualitative immunochromatographic lateral flow test cartridges designed for detection of allergenic food proteins
  • Designed to detect both processed and unprocessed allergen proteins
  • Test can accommodate environmental swab, CIP rinse water and food samples
Specifications
Format Kit
Product Type Fish Protein Rapid Kit
Test Type Allergen
For Use With (Application) Screening fish proteins in CIP final rinse water, environmental swab samples, food ingredients and processed food products
Quantity 25/Cs.