Learn More
American Research Products Inc Fish ThyroXine T4 ELISA Kit
Supplier:  American Research Products Inc CSBE08489F
Encompass
Fish ThyroXine T4 ELISA Kit from CUSABIO Detection Range 20 ng/mL-320 ng/mL Sensitivity 20 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29679
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.