Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Collagenase (From Clostridium histolyticum, Islet Isolation Grade)

Catalog No. BP26491 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP26491 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP26491

missing translation for 'validMainframeMsg'

  • Application: Collagenase is most suitable for pancreatic islet isolation
  • Unit Definition: One unit will liberate peptides from collagen equivalent in ninhydrin color to 3.0μmole of L-leucine in 18 hr at pH 7.4 and 37°C
  • Storage: 0° to 5°C, dessicate

TRUSTED_SUSTAINABILITY

Specifications

Color Yellowish Brown
pH 7.4
For Use With (Application) Suitable for pancreatic islet isolation
Content And Storage 0°C to 5°C
Source Clostridium histolyticum
Quantity 1 g
Product Type Collagenase
Molecular Weight (g/mol) 105000
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.