Learn More
Description
- Ladder is ideal for precise molecular weight determination of polypeptides in denaturing PAGE and Western blots
- Each protein contains an integral Strep-tag™ II sequence1 and may be detected on Western blots using Strep-Tactin™-AP conjugate2 or an antibody against Strep-tag II
- 50kDa protein band has greater intensity than other bands to serve as a reference
- Supplied in gel loading buffer
Specifications
Specifications
| Form | Liquid |
| Formulation | Supplied in gel-loading buffer |
| For Use With (Application) | Molecular weight determination of polypeptides analyzed in various Gel system |
| Kit Contents | <51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet |
| Quantity | 2 x 500 μL, 200 Loadings |
| Range | 10 to 200 kd |
| Storage Requirements | −20°C |
| Sufficient For | 200 Loadings |
| Product Type | Protein Molecular Weight Marker |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.