Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ exACTGene™ Lambda DNA Template

Catalog No. BP6115 Shop All Fisherbrand Products
Change view
Click to view available options
Lambda DNA Control Template, 100μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BP6115 Lambda DNA Control Template, 100μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP6115

missing translation for 'validMainframeMsg'

Optimized for routine PCR applications

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 ng/μL
Content And Storage -20°C
For Use With (Equipment) Fisher BioReagents™ exACTGene™ PCR kit, troubleshoot PCR parameters
Product Type Lambda DNA Template (1ng/μL)
Volume 100μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.