Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Microbiology Media: LB Broth, Lennox (Granulated)

Catalog No. p-4245114 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2000 g
5 kg
Packaging Type:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP9722-2 2000 g Poly Bottle
BP9722-500 500 g Poly Bottle
BP97225 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. BP9722-2
Only null left
Add to Cart
This item is not returnable. View return policy

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

  • Directions for Media Preparation: Add 20g to 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water) then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Before use, sterilize by autoclaving for 15 minutes.
  • Provides a nutrient-rich environment to enable faster growth of E. coli
  • Format: Granulated powder
TRUSTED_SUSTAINABILITY

Specifications

Form Granulated Powder
Media Type LB Broth, Lennox
Product Type Nutrient Broth
Formulation Casein Peptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L
For Use With (Application) Microbiology Media
Packaging Type Poly Bottle
Quantity 2000 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.