Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Fisher BioReagents™ Microbiology Media Additives: Acid Casein Peptone

Catalog No. p-4907857 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
Packaging Type:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP1424-100 100 g Poly Bottle
BP1424-2 2000 g Poly Pail
BP1424-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP1424-100
Only null left
Add to Cart

Additive for preparing bacterial nutrient media

Acid Casein Peptone

Dehydrated.

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Grade Molecular genetics
Media Type Acid Casein Peptone
Product Type Media Additive
For Use With (Application) Acid hydrolyzed casein's low concentrations of sodium chloride and iron permit toxin production
Packaging Type Poly Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.