Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Microbiology Media Additives: Tryptone

Catalog No. BP1421100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
2000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP1421-100 100 g
BP1421-500 500 g
BP1421-2 2000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP1421-100
  • $154.70
    $133.65 / Each

    Save $21.05

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $736.00
    $635.65 / Case of 6 EA

    Save $100.35

    14% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge

missing translation for 'validMainframeMsg'

Additive for preparing bacterial nutrient media

Note: This is a product of animal origin (pancreas).
TRUSTED_SUSTAINABILITY

Specifications

Form Dehydrated Powder
Grade Molecular genetics
Media Type Tryptone
Product Type Dehydrated Culture Media
For Use With (Application) Pancreatic digest of casein is used as a nitrogen source for bacteria
Packaging Type Poly Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.