Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ NZY Broth (Powder/Molecular Genetics)

Catalog No. BP2465500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging Type:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP2465500 500 g Poly Bottle
BP2465100 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP2465500

missing translation for 'validMainframeMsg'

Specifications

Content And Storage Keep refrigerated. (Store below 4°C/39°F.) Store in a cool, dry, well-ventilated area.
Form Powder
Grade Molecular genetics
Media Type NZY Broth
Product Type Microbiology Media
Packaging Type Poly Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.