Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ OptiGrow™ Preweighed Media

Catalog No. BP142610P1 Shop All Fisherbrand Products
Change view
Click to view available options
Form:
Solid
Product Type:
Dehydrated Culture Media
Nutrient Agar
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Form Product Type
BP142610P1 Solid Dehydrated Culture Media
BP142510P1 Solid Nutrient Agar
BP246810P1 Solid Dehydrated Culture Media
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP142610P1
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Optimized for superior yield of bacterial culture

Four classic media, in convenient, preweighed pouches.

  • Preblended growth-promoting components save time and preparation costs
  • Reduce formulation errors
  • Each pouch contains enough material for 1 liter of liquid media
  • No more weighing of dusty powders
  • High quality—assured by stringent testing procedures

  • LB is used for the culturing of E. coli in molecular biology procedures
  • Terrific Broth is a buffered enriched medium used to obtain high yields of plasmid DNA from transformed E. coli

Order Info

Supplied as 10 pouches per box.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 4°C
Form Solid
Grade Molecular genetics
Media Type LB Broth, Miller
Product Type Dehydrated Culture Media
For Use With (Application) Culturing of Escherichia coli
Packaging Type Pouch
Quantity 10 Pouches
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.