Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Recombinant DNase 10X RXN Buffer, Optizym™, Fisher BioReagents

Catalog No. BP32261 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP32261 1000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP32261

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for use in RT-PCR and other RNA applications

  • Efficiently hydrolyzes phosphodiester linkages of double- or single-stranded DNA
  • Requires magnesium and calcium for maximal activity
  • Tested for RNase activity, protease activity, and in specific performance tests
  • Storage Buffer Components: 20mM HEPES (pH 7.5), 10mM CaCl2, 10mM MgCl2, 1mM DTT, 50% (v/v) glycerol
  • Provided with 10X Reaction Buffer: 100mM Tris-HCl (pH 7.5), 25mM MgCl2, 5mM CaCl2
  • Unit Definition: One unit is defined as the amount of enzyme required to completely degrade 1μg DNA in 10 minutes at 37°C
  • Unit present in each tube: 1mL per ampule
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 U/μL
For Use With (Application) RT-PCR, other RNA applications
Storage Buffer 20mM HEPES (7.5 pH), 10mM CaCl2, 10mM MgCl2, 1mM DTT, 50% Glycerol
Content And Storage -20°C
Quantity 1000 U
Product Type OPTIZYME™ rDNase I
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.