Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Optizyme™ Ribonuclease Inhibitor, Recombinant

Catalog No. BP32221 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
2500 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP3222-1 2500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP3222-1

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protects RNA from degradation by RNases

  • Supplied as a recombinant product from E. coli (originally isolated from rat lung) and provides superior protection of RNA from degradation by RNases
  • Commonly added to all solutions used during the isolation of RNA, and will not interfere with the performance of most enzymes in downstream applications
  • Active over a broad temperature range, and even provides some RNase inhibition at 60°C, which is useful when performing reverse transcription reactions at elevated temperatures to overcome secondary structure in RNA
  • Storage Buffer Components: 20mM HEPES-KOH (pH 7.6 at 4°C), 50mM KCl, 8mM DTT, and 50% glycerol (v/v)

Inactivates RNase A, RNase B, and RNase T2; No activity against SP6, T7, or T3 RNA polymerases, AMV or MMLV reverse transcriptases, and Taq DNA polymerase

  • Displays a broad spectrum of inhibitory activity against RNases, and does not have activity against other polymerases and reverse transcriptases
  • Suitable for use in common molecular biology applications such as isolation and purification of RNA, cDNA synthesis, RT-PCR, in vitro RNA transcription/translation, ribonuclease protection assay, and preparation of RNase-free antibodies

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 to 40 U/μL
Color Colorless
For Use With (Application) Isolation and purification of RNA, cDNA synthesis, RT-PCR, in-vitro RNA transcription/translation, Ribonuclease protection assay and preparation of RNase-free Antibodies
Storage Buffer 20mM HEPES-KOH (pH 7.6 at 4°C), 50mM KCl, 8mM DTT, 50% Glycerol
Source E. coli
Quantity 2500 U
Product Type RNase Inhibitor
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.