Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Peptone (Granulated)

Catalog No. BP97252 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging Type:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP9725-2 2 kg Poly Bottle
BP9725-500 500 g Poly Bottle
BP97255 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP9725-2
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

  • Mixture of peptides and free amino acids from pancreatic hydrolysis of animal tissue
  • pH (at 25°C, 2% Solution): 6.5 to 7.5
  • CAS: 73049-73-7

TRUSTED_SUSTAINABILITY

Specifications

Form Granulated Powder
Grade Molecular genetics
Media Type Peptone
Product Type Media Additive
Formulation Total Nitrogen <10%, Amino Nitrogen <3.5%
Packaging Type Poly Bottle
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.