Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Thrombin, From Bovine Plasma

Catalog No. BP25432 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 kU
2 kU
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP25432 2 kU
BP254310 10 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP25432

missing translation for 'validMainframeMsg'

  • Application: Thrombin is used for clotting factor tests, coagulation assays, and the defibrination of blood or plasma
  • Storage: Store below 0°C

TRUSTED_SUSTAINABILITY

Specifications

Color White
For Use With (Application) Clotting factor tests, coagulation assays and the defibrination of Blood or Plasma
Content And Storage <0°C
Source Bovine Plasma
Quantity 2 kU
Product Type Thrombin
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.