Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Tryptone (Granulated)
GSA_VA

Catalog No. BP97265 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2 kg
5 kg
Packaging Type:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
BP97265 5 kg Poly Pail
BP9726-2 2 kg Poly Bottle
BP9726-500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP97265
  • $1,918.00
    $1,545.65 / Each of 1

    Save $372.35

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

An enzymatic digest of casein, a milk protein

  • pH (at 25°C, 2% Solution): 6.5 to 7.3

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Form Granulated Powder
Media Type Tryptone
Product Type Microbiology Media
Formulation Total Nitrogen ≥10%, Amino Nitrogen ≥3.5%
Packaging Type Poly Pail
Quantity 5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.