Learn More
Fisher Healthcare™ PROTOCOL™ Cytologic Fixative
Catalog No. 23291719 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50mL
12 x 4 oz.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-291719 12 x 4 oz.
23-245688 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-291719
This item is not returnable. View return policy
Water soluble film dries over slides, producing a tough protective covering
For fixation of cytology slides in preparation for storage or transport. Water soluble film dries over slides, producing a tough protective covering.
Specifications
Product Type Fixative
Quantity 12 x 4 oz.
For Use With (Application) Storage or transport