Learn More
Description
- Polypropylene
- Autoclavable
- Graduation marks clearly identify volume
- Empty containers come with adhesive labels for patient identification and hazardous chemical warning
Specifications
Specifications
| Graduated | Yes |
| Material | Polypropylene |
| Capacity (English) | 4 oz. |
| Capacity (Metric) | 120 mL |
| Includes | 300 Containers |
| Label | Adhesive |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.