Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ PROTOCOL™ Empty Containers

Catalog No. 22316158 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
20 mL
40 mL
60 mL
90 mL
120 mL
Includes:
200 Containers
300 Containers
400 Containers
500 Containers
600 Containers
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Includes
22-316158 40 mL 600 Containers
22-032061 20 mL 200 Containers
22-032062 120 mL 300 Containers
22-316160 90 mL 400 Containers
22-316159 60 mL 500 Containers
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-316158
This item is not returnable. View return policy

Eliminate formalin evaporation

  • Polypropylene
  • Autoclavable
  • Graduation marks clearly identify volume
  • Empty containers come with adhesive labels for patient identification and hazardous chemical warning

Specifications

Graduated Yes
Material Polypropylene
Capacity (English) 1.375 oz.
Capacity (Metric) 40 mL
Includes 600 Containers
Label Adhesive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.