Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Healthcare™ Replacement Bowls for Tissue Flotation Baths and Flotation Work Stations

For use with Fisher HealthCare Tissue Flotation Baths and Flotation Work Stations
$62.40
Specifications
For Use With (Equipment) | Tissue flotation bath and flotation work station |
---|---|
Type | Replacement Bowl |
Catalog Number | Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Size | Price | Quantity & Availability | ||||||
22-047-783
![]() |
8 x 11 x 2 in. |
Each for $62.40
|
|
||||||
22-047-782
![]() |
8 x 8 x 2 in. |
Each for $62.40
|
|
||||||
Specifications
Tissue flotation bath and flotation work station | |
Replacement Bowl |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title