Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ Color Staph ID Latex Test Kit
SDP

Catalog No. 23046501 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
150 Tests
300 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-046-501 300 Tests
23-046-500 150 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-046-501
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Latex particle reagent, positive and negative controls, control latex reagent, disposable white slide cards (8-well), disposable mixing sticks, and package insert

The Sure-Vue Color Staph is a rapid, color enhanced latex agglutination test to detect clumping factor and/or Protein-A, two characteristics associated with Staphylococcus aureus colonies obtained from culture.

  • CLIA Complexity: Non-Waived
  • CPT Code: 87077
  • Sensitivity: 99%
  • Specificity: 100%
  • Latex agglutination slide test for the qualitative detection of Staphylococcus aureus from primary cultures
  • Produces visible red agglutination in a blue background when results are positive
  • Results within 30 seconds
  • Unused reagents can safely be used up to expiration date stamped on the box with prompt recapping and refrigeration
  • For in vitro diagnostic use

Specifications

CE Marker Self-declared
Clia Complexity Moderate (Serum, Plasma or Whole Blood), Waived (Other)
Detectable Analytes S. aureus
Type Sure-Vue™ Color Staph ID Latex Test Kit
Disposable Single-use
Includes Latex particle reagent, positive and negative control, control Latex reagent, disposable white slide cards (8-well) and disposable mixing sticks
Quantity 300 Tests
Form Ready-to-Use (RTU)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.